Would you like to log in again?
Round Bottom, Half Jacketed, Three-Neck Flasks, Chemglass
Supplier: Chemglass
CG-1537-02 CG-1537-10 CG-1537-20 CG-1537-11 CG-1537-14 CG-1537-24 CG-1537-01 CG-1537-26 CG-1537-15 CG-1537-08 CG-1537-06 CG-1537-09 CG-1537-05 CG-1537-04 CG-1537-22 CG-1537-03
89028-604EA 981.86 USD InStock
89028-604 89028-608 89028-606 80061-308 80061-318 80061-300 80061-310 80061-302 80061-312 80061-304 80061-314 80061-306 80061-316 80061-298 89028-610 80061-320
Round Bottom, Half Jacketed, Three-Neck Flasks, Chemglass
Flasks Round Bottom Flasks
These round bottom three-neck flasks feature heavy wall construction have [ST] outer joints on the center neck and vertical side necks
Lower half of the flask is jacketed with two hose connections for circulation of heating or cooling liquid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty