Would you like to log in again?
Flasks, Dewar, Wide Mouth, Metal Housing, with Carrying Handle, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1595-02 CG-1595-03 CG-1595-01
80061-510EA 408.01 USD InStock
80061-510 80061-512 80061-514
Flasks, Dewar, Wide Mouth, Metal Housing, with Carrying Handle, Chemglass
Dewars and Cryogenic Vessels
Dewar flask with full length aluminum housing which extends to the top rim for added protection.
  • Features carrying handle
  • Polyethylene stopper minimizes evaporation
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty