Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Lower Support Rings for Chemglass Reaction Vessels, Chemglass
Supplier: Chemglass
CG-1948-03 CG-1948-02 CG-1948-01
80061-830EA 159.11 USD InStock
80061-830 80061-832 80061-834
Lower Support Rings for Chemglass Reaction Vessels, Chemglass
Support Rings
Support rings for reaction vessels.
  • Support ring is constructed of 1/2" O.D.
  • PTFE coated stainless steel rod
Made with an opening for easy insertion of reactor with an 8" long rod for connection to lattice.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty