Would you like to log in again?
Caps, GL Thread with Aperature, Chemglass
Supplier: Chemglass
CG-195-03 CG-195-04 CG-195-02 CG-195-01 CG-195-05
80061-840EA 4.79 USD InStock
80061-840 80061-842 80061-844 80061-836 80061-838
Caps, GL Thread with Aperature, Chemglass
Closures Bottle Closures
Cap is made from PBT (thermoplastic polyester). It can be combined with the sealing ring will accommodate tubing of the listed diameter.
  • Open top cap for use with GL screw threads
  • Can withstand temperatures from –45 to 200 °C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty