Would you like to log in again?
SILICONE SEALING RING GL #14
Supplier: Chemglass
EXTRACTOR BODY, MODULAR, LIQUID-LIQUID^ Replacement Components. PTFE faced sealing ring for GL-14 thread. Modular extractor body for use with plain or Snyder style sidearm. For use with a 1,000ml sample and 200ml of solvent. Longer chamber length increases solvent residence time and efficiency.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SILICONE SEALING RING GL #14 80061-954 CG-197-01 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty