Would you like to log in again?
Magnetic Stirring Bars, Octagonal,PTFE, Chemglass
Supplier: C-Line
CG-2001-08 CG-2001-03 CG-2001-50 CG-2001-15 CG-2001-02 CG-2001-20 CG-2001-04 CG-2001-14 CG-2001-13 CG-2001-07 CG-2001-18 CG-2001-09 CG-2001-05 CG-2001-06 CG-2001-01 CG-2001-17 CG-2001-12 CG-2001-19 CG-2001-10 CG-2001-11
80062-044EA 27.33 USD InStock
80062-044 80062-066 80062-054 80062-076 80062-042 80062-064 80062-052 80062-074 80062-040 80062-062 80062-050 80062-072 80062-060 80062-048 80062-058 80062-068 80062-046 80062-078 80062-056 89028-972
Magnetic Stirring Bars, Octagonal,PTFE, Chemglass
Stirrers Magnetic Stirring Bars
Octagon cross-section features a molded-in pivot ring
The PTFE coated Alnico magnet permits magnetic agitation without contamination.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty