Would you like to log in again?
STIRRER SHAFT 480MM OAL
Supplier: Chemglass
STIRRER SHAFT, 10mm POLISHED^ Shaft Length: 480mm. Precision ground and polished shaft for use with fluoropolymer resin bearings and CG-2091 and CG-2092 agitator blades. With a flat ground section half the listed length of the shaft to permit positive seating of fluoropolymer resin locking screws.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STIRRER SHAFT 480MM OAL 80062-304 CG-2075-B-02 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty