Would you like to log in again?
STIRRER BLADE 19MM PERF. 3L
Supplier: Chemglass
STIRRER BLADE, 19MM HEAVY DUTY, FLUOROPOLYMER RESIN, BANANA SHAPE^ Perforated Blades. Fits Flask Capacity: 3000ml; Minimum Joint Clearance: 34/45. Blade is shaped to resemble the contour of round bottom flasks. Made of 3/16in. fluoropolymer resin to fit 19mm O.D. stirrer shafts.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STIRRER BLADE 19MM PERF. 3L 80062-404 CG-2090-13 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty