Would you like to log in again?
STIRRER PADDLE PTFE 120MM
Supplier: Chemglass
STIRRER PADDLE, FLUOROPOLYMER RESIN, 10mm^ Diameter: 120mm. All fluoropolymer resin four bladed angled agitator for use with 10mm O.D. shaft (CG-2075-B). Shaft opening is drilled straight through to permit positioning of paddle at any height on shaft.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STIRRER PADDLE PTFE 120MM 80062-416 CG-2091-03 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty