Would you like to log in again?
Needles for Syringes, Chemglass
Supplier: Chemglass
CG-3075-42 CG-3075-41 CG-3075-01 CG-3075-02 CG-3075-40 CG-3075-03
80062-644EA 139.26 USD InStock
80062-644 80062-600 80062-642 80062-598 80062-596 80062-640
Needles for Syringes, Chemglass
Syringe Needles
All needles are made of #304 full hard stainless steel and have a 22º beveled point for septum penetration along with general use.
Delivery information: Removable needles come complete with the PTFE ferrule necessary for assembly.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty