Would you like to log in again?
J-KEM® Temperature Controller, Model 210, Chemglass
Supplier: Chemglass
CG-3200-01 CG-3200-02 CG-3200-03 CG-3200-100
80062-838EA 1806.85 USD InStock
80062-838 80062-836 80062-840 10034-456
J-KEM® Temperature Controller, Model 210, Chemglass
Controllers Temperature Controllers
Sufficient for heating mantles up to 5L in size along with most laboratory ovens and hot plates
Controller has the ability to display temperature without heating. The Model 210 controls temperature to within ± 0.1 °C of liquids from 1µL to 5L.
Ordering information: Includes 1/8” dia. x 12” long PTFE coated thermocouple, 10 feet coiled extension cord, and a 14/20 PTFE thermocouple adapter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty