Would you like to log in again?
T TEMP. CONTROLLER MODEL 26
Supplier: Chemglass
TEMPERATURE CONTROLLER, J-KEM MODEL260/T^ Complete System. Thermocouple Type: 'T'; Temperature Range: -200 to 250[degree]C. Dimensions: 3.25in. Highx7.5in. Widex9in. Deep. Power: 15amps@120VAC, 1800Watts. J-KEM Scientific microprocessor based temperature controller.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
T TEMP. CONTROLLER MODEL 26 80062-958 CG-3206-01 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty