Would you like to log in again?
O'RING KITS GLASS STEM 0-4 V
Supplier: Chemglass
O-RING REPLACEMENT KITS, GLASS STEM^ To Fit Plug Size: 0-4; O-Ring Type: Viton. Tip O-Ring Size: 007. Shaft O-Ring Size: 009. Each kit contains a set of three o-rings, one for the tip and two for the barrel seal. O-ring sizes are listed in the following Chart.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
O'RING KITS GLASS STEM 0-4 V 80063-238 CG-585-01 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty