Would you like to log in again?
Krytox® General Purpose Grease, Chemglass
Supplier: Chemglass
Krytox®
CG-GPL-205-01
80063-672EA 142.93 USD InStock
80063-672
Krytox® General Purpose Grease, Chemglass
Lubricants
Krytox® GPL general purpose grease for use with standard taper joints, stopcocks and stirrer bearings. Useful at temperatures up to 400 °F continuously and 550 ºF intermittently. Minimum useful temperature is –35 °F.
  • Highly inert
  • Non-flammable grease for use with standard taper joints and glass stopcocks
  • Insoluble in almost all solvents except Freon 113®
  • Supplied in convenient 2 oz. tubes
Krytox® greases have demonstrated an exceptionally high degree of chemical inertness to many highly reactive chemicals. It has shown no evidence of reactivity when exposed to: Boiling Sulfuric Acid, Fluorine Gas, Nitrogen, Tetroxide, Chlorine Triflouride, Hydrogen Gas, Methanol, Ammonia Gas, Ethyl Alcohol, Hydrochloric Acid, Hydrazine, Diethylenetriamine, Phosphoric Acid, Sulfuric Acid, Chlorine, or Hydrogen Peroxide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty