Would you like to log in again?
Krytox® Low Vapor Pressure Grease, Chemglass
Supplier: Chemglass
Krytox®
CG-LVP-01
80063-674EA 339 USD InStock
80063-674
Krytox® Low Vapor Pressure Grease, Chemglass
Lubricants
Krytox® LVP grease is made with low vapor pressure fluorinated oils and is designed for use in high vacuum applications down to -1.0×10-13 Torr at 20 °C, and -1.0×10-5 Torr at 200 °C. The useful temperature range of Krytox® LVP is -20 to 260 °C.
  • Highly inert
  • Non-flammable grease for use with standard taper joints and glass stopcocks
  • Insoluble in almost all solvents except Freon 113®
  • Supplied in convenient 2 oz. tubes
Krytox® greases have demonstrated an exceptionally high degree of chemical inertness to many highly reactive chemicals. It has shown no evidence of reactivity when exposed to: Boiling Sulfuric Acid, Fluorine Gas, Nitrogen, Tetroxide, Chlorine Triflouride, Hydrogen Gas, Methanol, Ammonia Gas, Ethyl Alcohol, Hydrochloric Acid, Hydrazine, Diethylenetriamine, Phosphoric Acid, Sulfuric Acid, Chlorine, or Hydrogen Peroxide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty