Would you like to log in again?
QTZ. TO BORO GRADED SEAL 4 X
Supplier: Chemglass
GRADED SEAL, QUARTZ TO BOROSILICATE^ O.D.xI.D; 4x6mm. Graded seals have 3in. of quartz sealed to 4in. of borosilicate.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
QTZ. TO BORO GRADED SEAL 4 X 80063-724 CGQ-0310-03 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty