Would you like to log in again?
Quartz Ground and Polished Plates, Chemglass
Supplier: Chemglass
CGQ-0620-14 CGQ-0620-10 CGQ-0620-06 CGQ-0620-04 CGQ-0620-05 CGQ-0620-19 CGQ-0620-09 CGQ-0620-02 CGQ-0620-15 CGQ-0620-01 CGQ-0620-07 CGQ-0620-21 CGQ-0620-17 CGQ-0620-03 CGQ-0620-12 CGQ-0620-20 CGQ-0620-11 CGQ-0620-18 CGQ-0620-16 CGQ-0620-08
80063-838EA 180.13 USD InStock
80063-838 80063-816 80063-848 80063-826 80063-818 80063-828 80063-812 80063-834 80063-822 80063-844 80063-814 80063-824 80063-846 80063-830 80063-852 80063-840 80063-832 80063-820 80063-842 80063-850
Quartz Ground and Polished Plates, Chemglass
Optical Windows
Ground and polished clear fused quartz discs with edges that have a ground finished on all sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty