Would you like to log in again?
QUARTZ ROD 8MM EA=4FT
Supplier: Chemglass
HERAEUS AMERSIL CLEAR FUSED HSQ-100 QUARTZ ROD^ O.D; 8mm; Tolerance O.D; +/-0.6mm. Commercial grade quartz rod. Stocked in 4 foot lengths. NOTE: PRICE LISTED IS PER FOOT. ROD IS NORMALLY STOCKED IN 4 FOOT LENGTHS.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
QUARTZ ROD 8MM EA=4FT 80063-990 CGQ-0801-07 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty