Would you like to log in again?
Micro Spatula, Chemglass
Supplier: Chemglass
MW-131-01
80064-042EA 24.35 USD InStock
80064-042
Micro Spatula, Chemglass
Spatulas
Nickel-stainless spatula with PTFE coated ends.
  • One end tapered to 0.8 mm Ø
  • The opposite end is rounded with 4 mm Ø
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty