Would you like to log in again?
Minum-Ware® Air Reflux Condenser, Chemglass
Supplier: Chemglass
MW-40-01
80064-116EA 74.39 USD InStock
80064-116
Minum-Ware® Air Reflux Condenser, Chemglass
Condensers
Condenser uses the surrounding air to dissipate the heat of the reaction during refluxing.
  • Features a top threaded outer joint and lower inner drip joint
  • All joints are 14/10
Delivery information: Supplied with open-top screw cap and Viton O-ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty