Would you like to log in again?
CONDENSER WATER COOLED JACK
Supplier: Chemglass
CONDENSER, JACKETED REFLUX, MINUM-WARE^ Liebig-type water jacketed condenser with a jacket length of 80mm. With top threaded outer and lower inner drip joint, both 14/10. Supplied complete with open-top screw cap (MW-102-33) and Viton O-ring (MW-100-08).
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CONDENSER WATER COOLED JACK 80064-118 MW-42-01 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty