Would you like to log in again?
CONDENSER COLD FINGER ONLY
Supplier: Chemglass
CONDENSER, COLD FINGER, ADJUSTABLE MINUM-WARE^ Cold Finger Only. Cold finger style condenser is made to fit into MW-50 Hickman still head while the lower drip tip directs distillate into the collection collar.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CONDENSER COLD FINGER ONLY 80064-122 MW-43-02 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty