Would you like to log in again?
FUNNEL COMPLETE MEDIUM
Supplier: Chemglass
FUNNEL, MICRO HIRSCH, MINUM-WARE^ Complete Item, Medium Porosity. Microscale vertically adjustable Hirsch funnel fo use with MINUM-WARE flasks or conical reaction vials having 14/10 joints. Funnel has a stem O.D. of 1/4in. and a length of 4in.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FUNNEL COMPLETE MEDIUM 80064-138 MW-53-01 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty