Would you like to log in again?
TUBE CRAIG COMP. 1ML
Supplier: Chemglass
RECRYSTALLIZATION TUBE, MINUM-WARE^ 1.0ml. Used for microscale recrystallizing of small quantities of reaction product. Consists of glass outer tube with non-uniform grind to purposely allow leakage, and fluoropolymer resin inner plug. Fluoropolymer resin inner plug fits both 1.0ml and 2.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE CRAIG COMP. 1ML 80064-198 MW-74-03 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty