Would you like to log in again?
Airfree® Schlenk Adapters, 15 mm O-Ring Joints, Chemglass
Supplier: Chemglass
Airfree®
AF-0070-02 AF-0070-07 AF-0070-10 AF-0070-09 AF-0070-04 AF-0070-20 AF-0070-11 AF-0070-08 AF-0070-06 AF-0070-05 AF-0070-01 AF-0070-03
80064-238EA 55.78 USD InStock
80064-238 80064-240 80064-250 80064-244 80064-254 89028-166 80064-242 80064-252 80064-248 80064-236 80064-246 80064-234
Airfree® Schlenk Adapters, 15 mm O-Ring Joints, Chemglass
Adapters Adapters, Joints with Tube Connectors
Adapters are designed for use with vacuum manifolds and permit the manifold to be utilized with a multitude of standard glassware or tubing products.
  • To be used with Cajon® fittings or at the end of manifolds, heavy-wall rubber vacuum tubing
  • Serrated hose connection adapters list the O.D. of the hose connection at the middle ring
  • Each adapter is supplied complete with one Viton O-Ring
Adapters for use with (80064-216;80064-218;80064-220), through (80064-228;80064-230;80064-232), vacuum manifolds permit the manifold to be utilized with a multitude of standard glassware or tubing products. The adapters are to be used with either a Cajon® fitting on the main ports or at the ends of the manifold with heavy-wall rubber vacuum tubing. Adapters are to be attached to the manifold via a (80073-354) S.S. pinch clamp.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty