Would you like to log in again?
Cajon® Airfree® Bubbler, Connection, Complete, Chemglass
Supplier: Chemglass
AF-0085-01
80064-292EA 271.32 USD InStock
80064-292
Cajon® Airfree® Bubbler, Connection, Complete, Chemglass
Manometers
Bubbler has an integral valve to control the flow of inert gas into the system.
Bubbler requires a minimal amount of oil due to the inner tube being within 5 mm of the bottom. For use with ³/₈" Cajon® fittings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty