Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Airfree® Schlenk Storage Vessels, with O-Ring, Chemglass

Supplier: Chemglass

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Airfree®
AF-0092-01 AF-0092-04 AF-0092-03 AF-0092-02
80064-318EA 292.06 USD InStock
80064-318 80064-316 80064-320 80064-322
Airfree® Schlenk Storage Vessels, with O-Ring, Chemglass
Reaction Vessels Reaction Vessels, Flask Shape
Storage vessel complete with an O-Ring joint on the sidearm.

  • #15 O-Ring joint
  • Chem-Vac™, Chem-Cap® valve to control gas or liquid flow into the vessel

Vessels are constructed of medium wall tubing and are used with manifolds or any other Airfree® component with a #15 O-Ring joint.

Delivery information: Supplied complete with one #116 Viton O-Ring.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty