Would you like to log in again?
Airfree Storage Flasks, Schlenk, Chemglass
Supplier: Chemglass
AF-0094-01 AF-0094-02 AF-0094-05 AF-0094-03 AF-0094-04
80064-338EA 259.11 USD InStock
80064-338 80064-342 80064-340 80064-336 80064-334
Airfree Storage Flasks, Schlenk, Chemglass
Flasks Round Bottom Flasks
Storage flask sidearm has a 9 mm OD tubulation for use with 3/8" Cajon® connections
Chem-Vac™ Chem-Cap® valve is used to control gas or liquid flow into the vessel.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty