Would you like to log in again?
Airfree® Schlenk Trap Connection Adapter, Chemglass
Supplier: Chemglass
Airfree®
AF-0300-14
80064-410EA 583.95 USD InStock
80064-410
Airfree® Schlenk Trap Connection Adapter, Chemglass
Adapters Adapters, Joints with Tube Connectors
Adapter used to connect the 80064-408 manifold to the 80064-400 trap assembly.
Adapter has a 0 to 20 mm Chem-Cap® valve with two #20 O-Ring joints.
Delivery information: Supplied complete with two #214 Viton O-Rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty