Would you like to log in again?
Airfree® Schlenk Connecting Adapters, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Airfree®
AF-0500-02 AF-0500-15 AF-0500-16 AF-0500-01
80064-470EA 203.44 USD InStock
80064-470 80064-474 80064-472 80064-476
Airfree® Schlenk Connecting Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
These adapters convert standard flasks and other apparatus to Airfree® applications.
  • Top outer and lower inner standard taper joints are same size
With top outer and lower inner standard taper joints, both being the same size.
Delivery information: Available with choice of 2 mm high vacuum glass stopcock and a stainless steel constant pressure retention device or 0 to 4 mm Chem-Cap® Airfree® inlet valve.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty