Would you like to log in again?
Airfree® Schlenk Adapter, Chemglass
Supplier: Chemglass
Airfree®
AF-0501-01 AF-0501-02
80064-480EA 233.93 USD InStock
80064-480 80064-478
Airfree® Schlenk Adapter, Chemglass
Adapters Adapters, Conical Ground Joints
Adapter allows standard single-neck round bottom flasks to be used as storage vessels..
  • Equipped with 0 to 4 mm Chem-Vac™ Chem-Cap® plug
Sampling can be accomplished by removing the 0 to 4 mm Chem-Vac™ Chem-Cap® plug, while maintaining inert gas purge through sidearm.
Ordering information: Available with a choice of 14/20 or 24/40 standard taper inner joint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty