Would you like to log in again?
Airfree® Schlenk Flushing Adapters, with Septum Stoppers, Chemglass
Supplier: Chemglass
Airfree®
AF-0508-20 AF-0508-21 AF-0508-10 AF-0508-11
80064-524EA 258.22 USD InStock
80064-524 80064-518 80064-522 80064-520
Airfree® Schlenk Flushing Adapters, with Septum Stoppers, Chemglass
Adapters Adapters, Conical Ground Joints
Adapter permits vessels to be purged with an inert gas during sampling or transfer with a cannulas.
  • Stopcocks have 9 mm O.D. medium wall arms
Both the glass and PTFE stopcock have 9 mm O.D. medium wall arms for use with red or white septum stopper. Additional arm is provided for connection to inert gas source.
Delivery information: Available with choice of 4 mm high vacuum glass stopcock or 4 mm PTFE stopcock. Supplied with two red septum stoppers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty