Would you like to log in again?
Airfree® Flushing Adapters, Chemglass
Supplier: Chemglass
Airfree®
AF-0509-10 AF-0509-11 AF-0509-13 AF-0509-12
80064-536EA 246.12 USD InStock
80064-536 80064-530 80064-534 80064-532
Airfree® Flushing Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
Flushing adapters allow vessels to be flushed with inert gas while sampling directly through the bore of the T-bore stopcock.
  • Two formats available
  • With glass stopcock or fluoropolymer resin plug
  • Septa not included
Available with a 4mm glass stopcock with constant pressure retention device or 4mm bore, 1–5 taper, fluoropolymer resin plug.
Ordering information: Not supplied with septa.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty