Would you like to log in again?
Airfree® Schlenk Flushing Adapters, without Septum Stoppers, Chemglass
Supplier: Chemglass
Airfree®
AF-0509-A-03 AF-0509-A-04 AF-0509-A-02 AF-0509-A-01
80064-538EA 265.12 USD InStock
80064-538 80064-540 80064-544 80064-542
Airfree® Schlenk Flushing Adapters, without Septum Stoppers, Chemglass
Adapters Adapters, Conical Ground Joints
Similar to flushing adapter but having a top outer joint and lower inner joint of the same size.
  • Stopcocks have 9 mm O.D. medium wall arms
  • Addition of the outer joint permits use of the larger sizes septum stoppers for repeated needle penetrations
Adapter also allows for evacuation and transfer of product into another vessel with the same size standard taper joints.
Ordering information: These adapters are not supplied with septum stopper.
Delivery information: Available with a choice of a 4 mm glass high vacuum stopcock with a stainless steel constant pressure retention device or 4 mm PTFE stopcock.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty