Would you like to log in again?
Airfree® Schlenk Horizontal Bubblers, Chemglass
Supplier: Chemglass
Airfree®
AF-0514-02 AF-0514-01
80064-570EA 355.27 USD InStock
80064-570 80064-572
Airfree® Schlenk Horizontal Bubblers, Chemglass
Bubblers
Horizontal bubblers manufactured by Chemglass.
  • Allows for in-line connection
  • With 9 mm O.D. middle serration
  • Equipped with standard taper inner joint
Horizontal design of this bubbler allows for in-line connection and purging of the apparatus. Hose connection has a 9 mm O.D. middle serration. Complete with standard taper inner joint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty