Would you like to log in again?
Airfree® Schlenk Storage Flasks with Sidearm, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Airfree®
AF-0522-08 AF-0522-06 AF-0522-01 AF-0522-07 AF-0522-03 AF-0522-02 AF-0522-05 AF-0522-04
80064-626EA 317.08 USD InStock
80064-626 80064-624 80064-618 80064-628 80064-622 80064-632 80064-620 80064-630
Airfree® Schlenk Storage Flasks with Sidearm, Chemglass
Reaction Vessels Reaction Vessels, Flask Shape
Round bottom storage flask with a standard taper outer joint on the sidearm.
The 50, 100 and 250 ml sizes have a 0 to 4 mm Chem-Vac™ Chem-Cap® valve, while the 500 and 1000 ml sizes have a 0 to 8 mm valve size.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty