Would you like to log in again?
Airfree® Reaction Flasks, Round Bottom, Schlenk, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AIRFREE®
AF-0528-23 AF-0528-05 AF-0528-26 AF-0528-25 AF-0528-04 AF-0528-01 AF-0528-22 AF-0528-21 AF-0528-20 AF-0528-03 AF-0528-24 AF-0528-02 AF-0528-06 AF-0528-07
80064-662EA 248.33 USD InStock
80064-662 80064-672 80064-650 80064-660 80064-670 80064-666 80064-654 80064-664 80064-674 80064-652 80064-648 80064-658 80064-668 80064-656
Airfree® Reaction Flasks, Round Bottom, Schlenk, Chemglass
Reaction Vessels Reaction Vessels, Flask Shape
These round bottom single neck flask having a standard taper outer joint.
Available with a choice of a 2 mm high vacuum glass stopcock having a S.S. constant pressure retaining device or a 0 to 4 mm Chem-Cap® Airfree® inlet valve.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty