Would you like to log in again?
Airfree® Schlenk Addition Funnels, Graduated, with Glass T-Bore Stopcock, Chemglass
Supplier: Chemglass
Airfree®
AF-0547-G-02 AF-0547-G-01
80064-912EA 509.17 USD InStock
80064-912 80064-914
Airfree® Schlenk Addition Funnels, Graduated, with Glass T-Bore Stopcock, Chemglass
Funnels Dropping Funnels
Pressure equalizing addition funnel, with graduations and a glass T-Bore stopcock in the equalizing arm to permit purging and isolation of the funnel contents for storage.
  • Glass T-Bore stopcock
  • Top and lower inner joints are the same size
  • 2 mm high vacuum glass stopcocks
Both top outer and lower inner joints are the same size. Lower stopcock is a glass straight bore stopcock. Both stopcocks are 2 mm high vacuum glass stopcocks with a stainless steel constant pressure retention device.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty