Would you like to log in again?
Airfree® Schlenk Addition Funnels, Chemglass
Supplier: Chemglass
Airfree®
AF-0548-20 AF-0548-01 AF-0548-21 AF-0548-02
80064-922EA 522.76 USD InStock
80064-922 80064-920 80064-916 80064-918
Airfree® Schlenk Addition Funnels, Chemglass
Funnels Dropping Funnels
Lower outlet is a standard taper inner joint with a 0 to 4 mm Chem-Vac™ Chem-Cap® valve.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty