Would you like to log in again?
Airfree® Schlenk Addition Funnels, Graduated, Chemglass
Supplier: Chemglass
AF-0551-01 AF-0551-02 AF-0551-03 AF-0551-04
80064-934EA 1028.72 USD InStock
80064-934 80064-932 80064-938 80064-936
Airfree® Schlenk Addition Funnels, Graduated, Chemglass
Arsine Generators
Unique design of this addition funnel permits solutions to be prepared using a magnetic stir bar.
  • Lower valve is a 0 to 4 mm Chem-Vac™ Chem-Cap® valve
  • 50 and 125 ml sizes are graduated in 1.0 ml subdivisions; 250 ml size is graduated in 5.0 ml subdivisions
Side arm has a 2 mm high vacuum glass T-Bore stopcock with a stainless steel constant pressure retension device for evacuation and purging both the funnel and lower vessel. Top outer and lower inner standard taper joints are the same size.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty