Would you like to log in again?
Straight Connecting Adapters, [ST] Inner Joints, Chemglass
Supplier: Chemglass
CG-1003-02 CG-1003-01
80065-584EA 90.88 USD InStock
80065-584 80065-586
Straight Connecting Adapters, [ST] Inner Joints, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters feature standard taper inner joints at both ends.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty