Would you like to log in again?
Combination Connecting Adapters, Top [ST] Joint, Lower Ball Joint, Chemglass
Supplier: Chemglass
CG-1006-01 CG-1006-03 CG-1006-02
80065-612EA 82.26 USD InStock
80065-612 80065-614 80065-616
Combination Connecting Adapters, Top [ST] Joint, Lower Ball Joint, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters feature a top standard taper outer joint and lower spherical ball joint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty