Would you like to log in again?
Inlet Adapters, [ST] Outer Joint, Chemglass
Supplier: Chemglass
CG-1013-02 CG-1013-01 CG-1013-14
80065-704EA 57.96 USD InStock
80065-704 80065-694 80065-696
Inlet Adapters, [ST] Outer Joint, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters allow for flexible tubing to be connected to apparatus for introduction of vacuum or pressure.
Hose connection has an O.D. of 10 mm at the largest serration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty