Would you like to log in again?
Claisen Distillation Adapters, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1015-14 CG-1015-19 CG-1015-01
80065-734EA 307.12 USD InStock
80065-734 80065-738 80065-740
Claisen Distillation Adapters, Chemglass
Distillation Components Distillation Adapters
Adapters feature a thermometer neck on the vertical side tube for immersion thermometers
The inner drip tip joint on the sidearm and the top outer joint and lower inner joint on the vertical tube are all the same [ST] joint size.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty