Would you like to log in again?
Claisen Adapters, with Thermometer Joint, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1025-01 CG-1025-02
80065-842EA 188.8 USD InStock
80065-842 80065-844
Claisen Adapters, with Thermometer Joint, Chemglass
Distillation Components Distillation Adapters
Adapters have a standard taper outer joint at the top of the main tube for use with jointed thermometers.
  • Sidearm has standard taper outer joint while main tube has an innter joint (both being the same size)
  • For use with the corresponding size jointed thermometers
  • Manufactured by Chemglass
The 10/18 size is used with 10/18, 25 mm immersion thermometers while the 10/30 is for use with CG-3506, 75 mm immersion thermometers. Sidearm has a standard taper outer joint with main tube having an inner joint, both being the same size.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty