Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Flow Control Adapters, 90°, with Glass Stopcock, Chemglass

Supplier: Chemglass

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CG-1028-02 CG-1028-29 CG-1028-01 CG-1028-19
80065-876EA 138.43 USD InStock
80065-876 80065-878 80065-866 80065-868
Flow Control Adapters, 90°, with Glass Stopcock, Chemglass
Adapters Adapters, Joints with Tube Connectors
Flow control adapters, used to regulate the flow of gases or liquids into reaction systems.

  • Supplied with a retaining clip
  • Equipped with 2 mm glass stopcock

Stopcock has a bore of 2 mm and is supplied complete with a retaining clip. Hose connection has an O.D. of 8 mm at the middle serration.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty