Would you like to log in again?
Flow Control Adapters, Straight, with PTFE Stopcock, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1029-19 CG-1029-29 CG-1029-01 CG-1029-14 CG-1029-02
80065-886EA 157.89 USD InStock
80065-886 80065-892 89028-204 80065-884 89028-202
Flow Control Adapters, Straight, with PTFE Stopcock, Chemglass
Adapters Adapters, Joints with Tube Connectors
Flow control adapters, used to regulate the flow of gases or liquids into reaction systems.
  • Equipped with PTFE stopcock
PTFE stopcock has a bore of 2 mm. Hose connection has an O.D. of 8 mm at the middle serration.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty