Would you like to log in again?
Flow Control Adapter, PTFE Metering Stopcock, Chemglass
Supplier: Chemglass
CG-1031-01
80065-922EA 274.12 USD InStock
80065-922
Flow Control Adapter, PTFE Metering Stopcock, Chemglass
Adapters Adapters, Joints with Tube Connectors
Adapter has a 2 mm PTFE metering stopcock between top outer and lower inner standard taper joints to permit flow control of fluids between apparatus.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty