Would you like to log in again?
Kjeldahl Distilling Trap Adapters, Same Size [ST] Joints, Chemglass
Supplier: Chemglass
CG-1032-02 CG-1032-01 CG-1032-14
80065-930EA 147.84 USD InStock
80065-930 80065-932 80065-928
Kjeldahl Distilling Trap Adapters, Same Size [ST] Joints, Chemglass
Distillation Components Distillation Adapters
Adapter is placed above flask to prevent carry-over of reactions which have a tendency to foam.
With top standard taper outer and lower inner joints, both being the same size.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty