Would you like to log in again?
Universal Inlet Adapters, Chemglass
Supplier: Chemglass
CG-1042-01 CG-1042-14
80065-980EA 68.57 USD InStock
80065-980 80065-972
Universal Inlet Adapters, Chemglass
Adapters Adapters, Joints with Tube Connectors
These universal adapters have a thread at the top for a compression cap and o-ring.
  • Universal
  • Threaded top
  • Inner [ST] joint
Adapters feature a #7 em-Thread at the top for compression cap and Viton® O-ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty